Tisk…

Cyclopentylamine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Aminocyclopentane

M-Clarity™ quality level = MQ100

C115002-100G C115002-25G
SIALC115002-100GEA 3580 CZK
SIALC115002-100G SIALC115002-25G
Cyclopentylamine, Sigma-Aldrich®
Cyclopentylamine
Vzorec: C₅H₁₁N
MW: 85.15
Bod varu: 108
Hustota: 0.8689 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001380
CAS číslo: 1003-03-8
EINECS: 213-697-3
UN: 3286
ADR: 3 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR