Tisk…

6-Bromoquinoline ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

H55111.03 H55111.06
H55111.03EA 1430 CZK
H55111.03 H55111.06
6-Bromoquinoline ≥97%
6-Bromoquinoline

Purity: 97% CAS No.: 5332-25-2 Molecular Formula: C9H6BrN Formula Weight: 208.06 Flash Point: 110°(230°F) Refractive Index: 1.6630 Melting Point: 24° MDL No.: MFCD00024023

Vzorec: C₉H₆BrN
MW: 208,06 g/mol
Bod tání: 24 °C
Hustota: 1,538
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00024023
CAS číslo: 5332-25-2
EINECS: 226-238-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR