2,2'-Dithiodianiline (thulium ionophore I) 97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2,2-Disulphanediyldianiline, 2,2'-Diaminodiphenyl disulphide, 2-Aminophenyl disulphide, Bis(2-aminophenyl) disulphide
H55807.03
H55807.06
H55807.14
H55807.03EA
755
CZK
H55807.03
H55807.06
H55807.14
2,2'-Dithiodianiline (thulium ionophore I) 97%
2,2'-Dithiodianiline (thulium ionophore I)
Purity: 97%
CAS No.: 1141-88-4
Molecular Formula: C12H12N2S2
Formula Weight: 248.36
Melting Point: 91-92°
Beilstein Reference: 914032
Vzorec:
C₁₂H₁₂N₂S₂ MW: 248,37 g/mol Bod tání: 91…92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007703 CAS číslo: 1141-88-4 EINECS: 214-529-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...