Triphenylphosphine oxide
Dodavatel: Cayman Chemical
9000289-25
9000289-50
9000289-500
9000289-1
CAYM9000289-25EA
675
CZK
CAYM9000289-25
CAYM9000289-50
CAYM9000289-500
CAYM9000289-1
Triphenylphosphine oxide
Triphenylphosphine oxide
Vzorec:
C₁₈H₁₅OP MW: 278.29 Bod varu: ∼360 °C (1013 hPa) Bod tání: 155…158 °C Hustota: 1.2 g/cm³ (20 °C) Bod vzplanutí: 180 |
MDL Number:
MFCD00002080 CAS číslo: 791-28-6 EINECS: 212-338-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...