Tisk…

3-Fluoro-5-iodoaniline 96%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

H34457.03 H34457.06
H34457.03EA 3400 CZK
H34457.03 H34457.06
3-Fluoro-5-iodoaniline 96%
3-Fluoro-5-iodoaniline

Purity: 96% CAS No.: 660-49-1 Molecular Formula: C6H5FIN Formula Weight: 237.02 MDL No.: MFCD04116466

Vzorec: C₆H₅FIN
MW: 237.02
Storage Temperature: Ambient
MDL Number: MFCD04116466
CAS číslo: 660-49-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR