2-Amino-9-fluorenone 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2-Amino-9H-fluoren-9-one
H55048.03
H55048.06
H55048.14
H55048.03EA
1790
CZK
H55048.03
H55048.06
H55048.14
2-Amino-9-fluorenone 98%
2-Amino-9-fluorenone
Purity: 98%
CAS No.: 3096-57-9
Molecular Formula: C13H9NO
Formula Weight: 195.22
Melting Point: 157-160°
MDL No.: MFCD00001157
Vzorec:
C₁₃H₉NO MW: 195,22 g/mol Bod varu: 361 °C (1013 hPa) Bod tání: 157…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001157 CAS číslo: 3096-57-9 EINECS: 221-445-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...