Tisk…

Methyl methanesulphonate ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

H55120.06 H55120.14 H55120.22
H55120.06EA 1970 CZK
H55120.06 H55120.14 H55120.22
Methyl methanesulphonate ≥99%
Methyl methanesulphonate

Purity: 99% CAS No.: 66-27-3 Molecular Formula: C2H6O3S Formula Weight: 110.13 Flash Point: 104°(219°F) Physical Form: Liquid Density: 1.3 Refractive Index: 1.4140 Appearance: Colourless to pale yellow Boiling Point: 202-203° Beilstein Reference: 1098586 Merck Reference: 14,6097 UN No.: 2810 MDL No.: MFCD00007557

Vzorec: C₂H₆O₃S
Bod varu: 202…203 °C
Bod tání: 20 °C
Hustota: 1,3
Bod vzplanutí: 104 °C (219 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007557
CAS číslo: 66-27-3
EINECS: 200-625-0
UN: 2810
ADR: 6.1,III
Merck Index: 14,6097

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR