4-Amino-2-methylpyridine 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
4-Amino-2-picoline, 2-Methyl-4-pyridylamine, 2-Methylpyridin-4-amine
H55036.MD
H55036.03
H55036.06
H55036.MDEA
1600
CZK
H55036.MD
H55036.03
H55036.06
4-Amino-2-methylpyridine 98%
4-Amino-2-methylpyridine
Purity: 98%
CAS No.: 18437-58-6
Molecular Formula: C6H8N2
Formula Weight: 108.14
Melting Point: 93-97°
MDL No.: MFCD00186509
Vzorec:
C₆H₈N₂ MW: 108,14 g/mol Bod tání: 93…97 °C Storage Temperature: Ambient |
MDL Number:
MFCD00186509 CAS číslo: 18437-58-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...