2-Bromo-4-chloropyridine ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
H33698.03
H33698.06
H33698.03EA
1260
CZK
H33698.03
H33698.06
2-Bromo-4-chloropyridine ≥97%
2-Bromo-4-chloropyridine
Purity: 97%
CAS No.: 22918-01-0
Molecular Formula: C5H3BrClN
Formula Weight: 192.44
Melting Point:
Vzorec:
C₅H₃BrClN Bod tání: <30 °C Storage Temperature: Ambient |
MDL Number:
MFCD03840757 CAS číslo: 22918-01-0 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...