Tisk…
C Columns, Empty Chromatography Columns
Dodavatel: Cytiva
19-5001-01 19-5003-01 19-5101-01 19-5102-01 19-5103-01 19-5104-01 19-5201-01 19-5202-01 19-5203-01 19-5002-01
19-5001-01EA 6780 CZK
19-5001-01 19-5003-01 19-5101-01 19-5102-01 19-5103-01 19-5104-01 19-5201-01 19-5202-01 19-5203-01 19-5002-01
C Columns, Empty Chromatography Columns
Kolony chromatografické
C columns are economical empty columns suitable for low pressure liquid chromatography and gravity feed at the lower volumes used at research scale.
  • Economical – for low pressure liquid chromatography at research scale
  • Broad compatibility – functions with a stand-alone pump, chromatography system, or gravity feed
  • Precise temperature control – with the optional cooling jacket
  • Efficient packing – packing reservoirs are available as accessories specific to each column diameter
Standard C-series columns have a fixed volume, but they can be upgradedMore Product Information