Tisk…
Alcohols, Sterile, Klercide™ 70/30 IPA Blended with WFI and Klercide™ 70/30 Denatured Ethanol Blended with WFI
Dodavatel: ECOLAB CONTAMINATION CONTROL
6600940 6600964 6600950
SHIE6600940EA 21540 CZK
SHIE6600940 SHIE6600964 SHIE6600950
Alcohols, Sterile, Klercide™ 70/30 IPA Blended with WFI and Klercide™ 70/30 Denatured Ethanol Blended with WFI
Prostředky čistící Dezinfekční prostředek
Contains 70% IPA or denatured ethanol (IMS) blended with highest specification (EP) water for injection. It is filtered to 0,2 µm, filled and triple bagged under ISO Class 5; Grade B (microbial count) laminar flow in an ISO Class 7; Grade C (microbial count) cleanroom before irradiation using a validated process at no less than 25 kGy to ensure spore free alcohol.
  • Endotoxin levels guaranteed to be below 0,25 EU/ml
  • Ideal for Grade A and B cleanrooms and product contact areas
  • SteriShield Delivery System™ (trigger spray) ensures 6 months in-use shelf life
  • Certificates of analysis, irradiation and sterility with every batch
  • Fully validated and CEN tested
Certifikace: EN standards: EN 1276 - Bactericidal activity; EN 1650 - Fungicidal activity; EN 13697 Bactericidal surface test.
Upozornení: Use biocides safely. Always read the label and product information before use