Tisk…
Decontamination solution, RNase AWAY®
Dodavatel: Thermo Fisher Scientific
7000 7002 7003 7005-11
732-2270EA 680 CZK
732-2270 732-2351 732-2271 732-2352
Decontamination solution, RNase AWAY®
Prostředky čistící Dekontaminanty
Ready-to-use solution for fast, easy elimination of RNase contamination from work surfaces, instruments, pipettes, glass and consumables.
  • Easy cleaning of various types of surfaces
  • Ideal for use on materials that are not autoclavable
  • Replaces time-consuming processes such as DEPC treatment
Ready-to-use solution for eliminating DNA contamination. For cleaning work surfaces, instruments, pipettes, thermal cyclers, etc.