2,5-Diphenyl-p-benzoquinone 96%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
2,5-Diphenylquinone, [1,1:4,1-Terphenyl]-2,5-dione
A15369.14
A15369.06
A15369.14EA
3950
CZK
A15369.14
A15369.06
2,5-Diphenyl-p-benzoquinone 96%
2,5-Diphenyl-p-benzoquinone
Purity: 96%
CAS No.: 844-51-9
Molecular Formula: C18H12O2
Formula Weight: 260.30
Melting Point: 215-219°
Beilstein Reference: 2051812
Vzorec:
C₁₈H₁₂O₂ MW: 260.29 Bod tání: 215…219 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001600 CAS číslo: 844-51-9 EINECS: 212-680-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...