5-Methylbenzo[b]thiophene ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
A12666.03
A12666.06
A12666.03EA
2640
CZK
A12666.03
A12666.06
5-Methylbenzo[b]thiophene ≥97%
5-Methylbenzo[b]thiophene
Purity: 97%
CAS No.: 14315-14-1
Molecular Formula: C9H8S
Formula Weight: 148.23
Melting Point: 32-36°
Boiling Point: 71-73°/0.6mm
MDL No.: MFCD00052509
Vzorec:
C₉H₈S Bod varu: 71…73 °C (0,6 mmHg) Bod tání: 32…36 °C Hustota: 1,11 Storage Temperature: Ambient |
MDL Number:
MFCD00052509 CAS číslo: 14315-14-1 EINECS: 238-256-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...