3-Methylbenzo[b]thiophene, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
3-Methylbenzothiophene
M-Clarity™ quality level = MQ100
638587-1G
638587-5G
SIAL638587-1GEA
2430
CZK
SIAL638587-1G
SIAL638587-5G
3-Methylbenzo[b]thiophene, Sigma-Aldrich®
3-Methylbenzo[b]thiophene
Vzorec:
C₉H₈S MW: 148,23 g/mol |
MDL Number:
MFCD00040243 CAS číslo: 1455-18-1 EINECS: 215-934-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...