3-Methylbenzo[b]thiophene ≥98%
Dodavatel: Thermo Scientific
Synonyms:
3-Methylbenzothiophene
A15772.03
A15772.06
A15772.14
A15772.03EA
1590
CZK
A15772.03
A15772.06
A15772.14
3-Methylbenzo[b]thiophene ≥98%
3-Methylbenzo[b]thiophene
Purity: 98%
CAS No.: 1455-18-1
Molecular Formula: C9H8S
Formula Weight: 148.23
Flash Point: 104°(219°F)
Refractive Index: 1.6240
Melting Point: -40°
Boiling Point: 72-74°/2mm
Beilstein Reference: 111282
MDL No.: MFCD00040243
Vzorec:
C₉H₈S MW: 148,23 g/mol Bod varu: 72…74 °C (2 mmHg) Bod tání: –40 °C Hustota: 1,13 Bod vzplanutí: 104 °C (219 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040243 CAS číslo: 1455-18-1 EINECS: 215-934-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...