2-Methylbenzo[b]thiophene ≥98%
Dodavatel: Thermo Scientific
H32224.03
H32224.06
H32224.03EA
3330
CZK
H32224.03
H32224.06
2-Methylbenzo[b]thiophene ≥98%
2-Methylbenzo[b]thiophene
Purity: 98%
CAS No.: 1195-14-8
Molecular Formula: C9H8S
Formula Weight: 148.23
Melting Point: 51-52°
MDL No.: MFCD00216250
Vzorec:
C₉H₈S MW: 148,23 g/mol Bod tání: 51…52 °C Storage Temperature: Ambient |
MDL Number:
MFCD00216250 CAS číslo: 1195-14-8 EINECS: 214-792-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...