2,1,3-Benzothiadiazole, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
B10900-100G
B10900-25G
B10900-5G
SIALB10900-100GEA
16840
CZK
SIALB10900-100G
SIALB10900-25G
SIALB10900-5G
2,1,3-Benzothiadiazole, Sigma-Aldrich®
2,1,3-Benzothiadiazole
Vzorec:
C₆H₄N₂S MW: 136,18 g/mol |
MDL Number:
MFCD00005809 CAS číslo: 273-13-2 EINECS: 205-985-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...