Tisk…

1,3-Diethynylbenzene, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

632104-1G 632104-5G
SIAL632104-1GEA 1240 CZK
SIAL632104-1G SIAL632104-5G
1,3-Diethynylbenzene, Sigma-Aldrich®
1,3-Diethynylbenzene
Vzorec: C₁₀H₆
MW: 126,16 g/mol
MDL Number: MFCD00041686
CAS číslo: 1785-61-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR