Bismuth subsalicylate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
480789-100G
480789-500G
SIAL480789-100GEA
1080
CZK
SIAL480789-100G
SIAL480789-500G
Bismuth subsalicylate, Sigma-Aldrich®
Bismuth subsalicylate
Vzorec:
C₇H₅BiO₄ MW: 362,09 g/mol |
MDL Number:
MFCD00085368 CAS číslo: 14882-18-9 EINECS: 238-953-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...