4,4-Dimethyl-2-oxazoline ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
A12670.06
A12670.14
A12670.22
A12670.06EA
1730
CZK
A12670.06
A12670.14
A12670.22
4,4-Dimethyl-2-oxazoline ≥98%
4,4-Dimethyl-2-oxazoline
Purity: 98%
CAS No.: 30093-99-3
Molecular Formula: C5H9NO
Formula Weight: 99.13
Flash Point: 1°(33°F)
Refractive Index: 1.4190
Boiling Point: 99-100°
Beilstein Reference: 1209606
UN No.: 1993
MDL No.: MFCD00014507
Vzorec:
C₅H₉NO Bod varu: 99…100 °C Hustota: 0,940 Bod vzplanutí: 1 °C (33 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014507 CAS číslo: 30093-99-3 EINECS: 250-045-7 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...