9-Fluorenylmethyl chloroformate (Fmoc chloride), Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
(9H-Fluoren-9-yl)methyl carbonochloridate, 9-Fluorenylmethyl chloroformate, Fmoc-Cl, 9H-Fluoren-9-ylmethyl chloroformate, Chloroformic acid 9-fluorenylmethyl ester, FMOCCl
M-Clarity™ quality level = MQ100
23184-1G
23184-5G
SIAL23184-1GEA
5060
CZK
SIAL23184-1G
SIAL23184-5G
9-Fluorenylmethyl chloroformate (Fmoc chloride), Sigma-Aldrich®
9-Fluorenylmethyl chloroformate (Fmoc chloride)
Vzorec:
C₁₅H₁₁ClO₂ MW: 258,7 g/mol |
MDL Number:
MFCD00001138 CAS číslo: 28920-43-6 EINECS: 249-313-6 UN: 2923 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...