Tisk…

(-)-Shikimic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (3R,4S,5R)-3,4,5-Trihydroxycyclohex-1-ene-1-carboxylic acid, Shikimic acid

M-Clarity™ quality level = MQ200

S5375-10MG S5375-1G S5375-5G
SIALS5375-10MGEA 1350 CZK
SIALS5375-10MG SIALS5375-1G SIALS5375-5G
(-)-Shikimic acid, Sigma-Aldrich®
(-)-Shikimic acid
Vzorec: C₇H₁₀O₅
MW: 174,15 g/mol
MDL Number: MFCD00066278
CAS číslo: 138-59-0
EINECS: 205-334-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR