Tisk…

(-)-Shikimic acid ≥98%, off-white powder

Dodavatel: MP Biomedicals

Synonyms: (3R,4S,5R)-3,4,5-Trihydroxycyclohex-1-ene-1-carboxylic acid, Shikimic acid

02152058.1 02152058.5 0215205801 0215205805
ICNA02152058.1EA 1140 CZK
ICNA02152058.1 ICNA02152058.5 ICNA0215205801 ICNA0215205805
(-)-Shikimic acid ≥98%, off-white powder
(-)-Shikimic acid

Shikimic acid, more commonly known as its anionic form shikimate, is an important biochemical metabolite in plants and microorganisms. Shikimic acid is a precursor for the aromatic amino acids phenylalanine and tyrosine, indole, indole derivatives and aromatic amino acid tryptophan, many alkaloids and other aromatic metabolites, phenylpropanoids, flavonoids, tannins, and lignin.


A tri-hydroxy cyclohexene carboxylic acid important in biosynthesis of so many compounds that the shikimate pathway (Seven step metabolic route used by bacteria, fungi, algae, parasites and plants for the biosynthesis of aromatic amino acids (phenylalanine, tyrosine, and tryptophan)). is named after it.


Solubility (50 mg/ml H2O): clear to slightly hazy

Vzorec: C₇H₁₀O₅
MW: 174,15 g/mol
Bod tání: 182…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00066278
CAS číslo: 138-59-0
EINECS: 205-334-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR