(-)-Shikimic acid 98%
Dodavatel: Thermo Fisher Scientific
132700010
132700050
ACRO132700010EA
3140
CZK
ACRO132700010
ACRO132700050
(-)-Shikimic acid 98%
(-)-Shikimic acid
Vzorec:
C₇H₁₀O₅ Bod tání: 185…191 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066278 CAS číslo: 138-59-0 EINECS: 205-334-2 |
Výsledky specifikačního testu
Appearance | White to light beige Powder |
Infrared spectrum | Conforms |
Melting point | 183 to 191 °C |
Specific optical rotation | -170° to -185° (20 °C, 589 nm) (C=2, H2O) |
Titration with NaOH | 97.5 to 102.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...