(R)-(-)-Thiazolidine-4-carboxylic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
H-Thz-OH, L(-)-Thiazolidine-4-carboxylic acid
M-Clarity™ quality level = MQ200
T27502-100G
T27502-10G
SIALT27502-100GEA
7480
CZK
SIALT27502-100G
SIALT27502-10G
(R)-(-)-Thiazolidine-4-carboxylic acid, Sigma-Aldrich®
(R)-(-)-Thiazolidine-4-carboxylic acid
Vzorec:
C₄H₇NO₂S MW: 133.17 |
MDL Number:
MFCD00005212 CAS číslo: 34592-47-7 EINECS: 252-106-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...