tert-Butyl hydroperoxide ≥70%, Luperox® TBH70X, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ200
458139-100ML
458139-1L
458139-25ML
458139-4L
458139-18L
SIAL458139-100MLEA
2310
CZK
SIAL458139-100ML
SIAL458139-1L
SIAL458139-25ML
SIAL458139-4L
SIAL458139-18L
tert-Butyl hydroperoxide ≥70%, Luperox® TBH70X, Sigma-Aldrich®
tert-Butyl hydroperoxide
Vzorec:
C₄H₁₀O₂ Bod varu: 96,2 °C Hustota: 0,93 g/mL (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002130 CAS číslo: 75-91-2 EINECS: 200-915-7 UN: 3109 ADR: 5.2, |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...