Tisk…

Methylstyrene (mixture of isomers) stabilizovaný, Sigma-Aldrich®

Dodavatel: Merck


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)
M-Clarity™ quality level = MQ200

522864-1L 522864-250ML
SIAL522864-1LEA 2670 CZK
SIAL522864-1L SIAL522864-250ML
Methylstyrene (mixture of isomers) stabilizovaný, Sigma-Aldrich®
Methylstyrene (mixture of isomers)
Vzorec: C₉H₁₀
MW: 118,18 g/mol
MDL Number: MFCD00168066
CAS číslo: 25013-15-4
EINECS: 246-562-2
UN: 2618
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR