Tisk…

5-Methylthiophene-2-carboxaldehyde, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 5-Methylthiophene-2-Carbaldehyde, 5-Methyl-2-thiophenecarboxaldehyde, 5-Methylthiophene-2-carbaldehyde

M-Clarity™ quality level = MQ300

W320900-100G-K W320900-1KG-K
SIALW320900-100G-KEA 7410 CZK
SIALW320900-100G-K SIALW320900-1KG-K
5-Methylthiophene-2-carboxaldehyde, Sigma-Aldrich®
5-Methylthiophene-2-carboxaldehyde
Vzorec: C₆H₆OS
MW: 126,18 g/mol
MDL Number: MFCD00005434
CAS číslo: 13679-70-4
EINECS: 237-178-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR