Thiophene-2-carbonyl chloride ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
A14641.14
A14641.06
A14641.22
A14641.36
A14641.14EA
1190
CZK
A14641.14
A14641.06
A14641.22
A14641.36
Thiophene-2-carbonyl chloride ≥98%
Thiophene-2-carbonyl chloride
Purity: 98%
CAS No.: 5271-67-0
Molecular Formula: C5H3ClOS
Formula Weight: 146.60
Storage Temperature: 2°C to 8°C
Flash Point: 90°(194°F)
Refractive Index: 1.5900
Boiling Point: 105°/30mm
Beilstein Reference: 110145
UN No.: 3265
MDL No.: MFCD00005428
Vzorec:
C₅H₃ClOS Bod varu: 105 °C (30 mmHg) Hustota: 1,372 Bod vzplanutí: 90 °C (194 °F) |
MDL Number:
MFCD00005428 CAS číslo: 5271-67-0 EINECS: 226-092-4 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...