Bismuth antimonide 99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A17014.09
A17014.18
A17014.09EA
2390
CZK
A17014.09
A17014.18
Bismuth antimonide 99%
Bismuth antimonide
Purity: 99.99%
CAS No.: 12323-19-2
Molecular Formula: BiSb
Formula Weight: 330.73
UN No.: 1549
MDL No.: MFCD00014196
Vzorec:
BiSb Storage Temperature: Ambient |
MDL Number:
MFCD00014196 CAS číslo: 12323-19-2 UN: 1549 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...