Lithium tetrachlorocuprate 0.1 mol/l in THF, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Dilithium tetrachlorocuprate
M-Clarity™ quality level = MQ100
224308-100ML
224308-800ML
SIAL224308-100MLEA
2990
CZK
SIAL224308-100ML
SIAL224308-800ML
Lithium tetrachlorocuprate 0.1 mol/l in THF, Sigma-Aldrich®
Lithium tetrachlorocuprate
Vzorec:
Cl₄CuLi₂ MW: 219.24 g/mol |
MDL Number:
MFCD00011081 CAS číslo: 15489-27-7 UN: 2056 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...