1,3-Phenylenediacetic acid ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2,2'-(1,3-Phenylene)diacetic acid
A11429.14
A11429.04
A11429.09
A11429.14EA
8650
CZK
A11429.14
A11429.04
A11429.09
1,3-Phenylenediacetic acid ≥97%
1,3-Phenylenediacetic acid
Purity: 97%
CAS No.: 19806-17-8
Molecular Formula: C10H10O4
Formula Weight: 194.19
Odour: Odourless
Melting Point: 172-178°
Beilstein Reference: 2616749
MDL No.: MFCD00004341
Vzorec:
C₁₀H₁₀O₄ MW: 194,19 g/mol Bod tání: 172…178 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004341 CAS číslo: 19806-17-8 EINECS: 243-332-3 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...