Benzeneseleninic anhydride 98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Benzeneseleninic acid anhydride
199710050
199710250
ACRO199710050EA
4600
CZK
ACRO199710050
ACRO199710250
Benzeneseleninic anhydride 98%
Benzeneseleninic anhydride
Vzorec:
C₁₂H₁₀O₃Se₂ MW: 360,13 g/mol Bod tání: 170…173 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001991 CAS číslo: 17697-12-0 EINECS: 241-701-3 UN: 3283 ADR: 6.1,II |
Výsledky specifikačního testu
Appearance | White to yellow to light beige Crystalline powder or needles |
Infrared spectrum | Conforms |
Titration with NaOH | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...