Tisk…

Pyromellitic dianhydride, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

412287-500G 412287-100G
SIAL412287-500GEA 6860 CZK
SIAL412287-500G SIAL412287-100G
Pyromellitic dianhydride, Sigma-Aldrich®
Pyromellitic dianhydride
Vzorec: C₁₀H₂O₆
MDL Number: MFCD00005005
CAS číslo: 89-32-7
EINECS: 201-898-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR