2,4-Diaminophenol dihydrochloride ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
4-Hydroxy-1,3-Phenylenediammonium Dichloride, 2,4-Diaminophenol Dihydrochloride, Amidol
A17697.14
A17697.22
A17697.14EA
1230
CZK
A17697.14
A17697.22
2,4-Diaminophenol dihydrochloride ≥98%
2,4-Diaminophenol dihydrochloride
Purity: 98+%
CAS No.: 137-09-7
Molecular Formula: C6H8N2O.2HCl
Formula Weight: 197.07
Melting Point: ca 280° dec.
Beilstein Reference: 3913728
Merck Reference: 14,2983
UN No.: 2811
MDL No.: MFCD00012979
Vzorec:
C₆H₁₀Cl₂N₂O MW: 197,06 g/mol Bod tání: ∼ 280 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00012979 CAS číslo: 137-09-7 EINECS: 205-279-4 UN: 2811 ADR: 6.1,III Merck Index: 14,2983 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...