Tisk…

Trimellitic anhydride, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

B4600-1KG B4600-25G
SIALB4600-1KGEA 4160 CZK
SIALB4600-1KG SIALB4600-25G
Trimellitic anhydride, Sigma-Aldrich®
Trimellitic anhydride
Vzorec: C₉H₄O₅
MDL Number: MFCD00005925
CAS číslo: 552-30-7
EINECS: 209-008-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR