Trimellitic anhydride chloride ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
1,3-Dioxo-1,3-dihydroisobenzofuran-5-carbonyl chloride, 1,2,4-Benzenetricarboxylic acid 1,2-anhydride 4-chloride
L14382.14
L14382.22
L14382.14EA
1090
CZK
L14382.14
L14382.22
Trimellitic anhydride chloride ≥98%
Trimellitic anhydride chloride
Purity: 98%
CAS No.: 1204-28-0
Molecular Formula: C9H3ClO4
Formula Weight: 210.57
Storage Temperature: 2°C to 8°C
Melting Point: 67-69°
Beilstein Reference: 610823
UN No.: 3261
MDL No.: MFCD00005924
Vzorec:
C₉H₃ClO₄ Bod tání: 67…69 °C |
MDL Number:
MFCD00005924 CAS číslo: 1204-28-0 EINECS: 214-874-8 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...