Tisk…

Triclosan, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 5-Chloro-2-(2,4-dichlorophenoxy)phenol, 5-Chloro-2-(2, 4-dichlorophenoxy) phenol

72779-25G-F 72779-5G-F
SIAL72779-25G-FEA 3960 CZK
SIAL72779-25G-F SIAL72779-5G-F
Triclosan, Sigma-Aldrich®
Triclosan
Vzorec: C₁₂H₇Cl₃O₂
MW: 289,54 g/mol
MDL Number: MFCD00800992
CAS číslo: 3380-34-5
EINECS: 222-182-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR