Tisk…

4-(4-Chlorophenoxy)phenol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Chloro-4'-hydroxydiphenyl ether, 4-(4-Chlorophenoxy)phenol

M-Clarity™ quality level = MQ100

695343-25G 695343-5G
SIAL695343-25GEA 9670 CZK
SIAL695343-25G SIAL695343-5G
4-(4-Chlorophenoxy)phenol, Sigma-Aldrich®
4-(4-Chlorophenoxy)phenol
Vzorec: C₁₂H₉ClO₂
MW: 220,65 g/mol
MDL Number: MFCD00125839
CAS číslo: 21567-18-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR