Tisk…

Bis(methylthio)methane, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ300

W387800-100G-K W387800-1KG-K W387800-2.5KG-
SIALW387800-100G-KEA 4160 CZK
SIALW387800-100G-K SIALW387800-1KG-K SIALW387800-2.5KG-
Bis(methylthio)methane, Sigma-Aldrich®
Bis(methylthio)methane
Vzorec: C₃H₈S₂
MW: 108.23
MDL Number: MFCD00008564
CAS číslo: 1618-26-4
EINECS: 216-577-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR