Tisk…

Bismuth(III) citrate, Sigma-Aldrich®

Dodavatel: Merck
Varování


M-Clarity™ quality level = MQ100

480746-100G 480746-500G
SIAL480746-100GEA 1390 CZK
SIAL480746-100G SIAL480746-500G
Bismuth(III) citrate, Sigma-Aldrich®
Bismuth(III) citrate
Vzorec: C₆H₅BiO₇
Storage Temperature: Ambient
MDL Number: MFCD00050817
CAS číslo: 813-93-4
EINECS: 212-390-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR