Tisk…

Sodium mercaptoacetate (sodium thioglycolate) 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

148890500 148890010 148892500
ACRO148890500EA 2430 CZK
ACRO148890500 ACRO148890010 ACRO148892500
Sodium mercaptoacetate (sodium thioglycolate) 98%
Sodium mercaptoacetate (sodium thioglycolate)
Vzorec: HSCH₂COONa
Bod tání: >300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00043386
CAS číslo: 367-51-1
EINECS: 206-696-4
UN: 2923
ADR: 8,III

Objednat

Výsledky specifikačního testu

Appearance White to off-white to light pink Powder
Infrared spectrum Conforms
Titration Iodimetric ≥97.5 %
Elemental Analysis 11.41 to 11.99 % (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR