Tisk…

O-Phenyl chlorothioformate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Phenyl chlorothionocarbonate, Phenyl chlorothionoformate, O-Phenyl carbonochloridothioate

M-Clarity™ quality level = MQ200

234524-1G 234524-5G
SIAL234524-1GEA 915 CZK
SIAL234524-1G SIAL234524-5G
O-Phenyl chlorothioformate, Sigma-Aldrich®
O-Phenyl chlorothioformate
Vzorec: C₇H₅ClOS
MW: 172,63 g/mol
MDL Number: MFCD00004920
CAS číslo: 1005-56-7
EINECS: 213-736-4
UN: 3265
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR