Tisk…

O-Phenyl chlorothioformate 99%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Phenyl chlorothionocarbonate, Phenyl chlorothionoformate, O-Phenyl carbonochloridothioate

215490050 215490010 215490250
ACRO215490050EA 3300 CZK
ACRO215490050 ACRO215490010 ACRO215490250
O-Phenyl chlorothioformate 99%
O-Phenyl chlorothioformate
Vzorec: C₇H₅ClOS
MW: 172,63 g/mol
Bod varu: 81…83 °C (6 mmHg)
Hustota: 1,248 g/cm³
Bod vzplanutí: 81 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004920
CAS číslo: 1005-56-7
EINECS: 213-736-4
UN: 3265
ADR: 8,II

Objednat

Výsledky specifikačního testu

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5800 to 1.5830 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR