Tisk…

O-p-Tolyl chlorothioformate ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: O-4-Methylphenyl chlorothioformate, p-Tolyl chlorothionoformate

B23501.09 B23501.18
B23501.09EA 2450 CZK
B23501.09 B23501.18
O-p-Tolyl chlorothioformate ≥97%
O-p-Tolyl chlorothioformate

Purity: 97% CAS No.: 937-63-3 Molecular Formula: C8H7ClOS Formula Weight: 186.66 Storage Temperature: 2°C to 8°C Flash Point: 96°(204°F) Refractive Index: 1.5740 Boiling Point: 70-72°/1mm Beilstein Reference: 1448986 UN No.: 3265 MDL No.: MFCD00014466

Vzorec: C₈H₇ClOS
MW: 186,66 g/mol
Bod varu: 70…72 °C (1 mmHg)
Hustota: 1,210
Bod vzplanutí: 96 °C (204 °F)
MDL Number: MFCD00014466
CAS číslo: 937-63-3
EINECS: 213-333-3
UN: 3265
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR