(3-Acryloxypropyl)trimethoxysilane ≥94% stabilizovaný
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Stabilization:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
L16400.06
L16400.14
L16400.06EA
3180
CZK
L16400.06
L16400.14
(3-Acryloxypropyl)trimethoxysilane ≥94% stabilizovaný
(3-Acryloxypropyl)trimethoxysilane
Stabilized with 100 ppm BHT
Purity: 94%
CAS No.: 4369-14-6
Molecular Formula: C9H18O5Si
Formula Weight: 234.33
Flash Point: 123°(253°F)
Refractive Index: 1.4290
Boiling Point: 95-100°/3mm
Beilstein Reference: 2046320
UN No.: 1760
MDL No.: MFCD00054803
Vzorec:
C₉H₁₈O₅Si Bod varu: 95…100 °C (3 mmHg) Hustota: 1,059 Bod vzplanutí: 123 °C (253 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00054803 CAS číslo: 4369-14-6 EINECS: 419-560-6 UN: 1760 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...