Tisk…
Lithium-tert-butanolate 1 mol/l in THF, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
398195-250ML 398195-50ML
SIAL398195-250MLEA 5640 CZK
SIAL398195-250ML SIAL398195-50ML
Lithium-tert-butanolate 1 mol/l in THF, Sigma-Aldrich®
Lithium-tert-butanolate
Vzorec: C₄H₉LiO
MW: 80.06
MDL Number: MFCD00050479
CAS číslo: 1907-33-1
EINECS: 217-611-5