Tisk…

Lithium-tert-butanolate 1 mol/l in hexane, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

398209-250ML 398209-50ML
SIAL398209-250MLEA 6020 CZK
SIAL398209-250ML SIAL398209-50ML
Lithium-tert-butanolate 1 mol/l in hexane, Sigma-Aldrich®
Lithium tert-butanolate
Vzorec: C₄H₉LiO
MW: 80,06 g/mol
MDL Number: MFCD00050479
CAS číslo: 1907-33-1
EINECS: 217-611-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR