Tisk…

DL-Ephedrine hydrochloride, Sigma-Aldrich®

Dodavatel: Merck
Varování


M-Clarity™ quality level = MQ100

285749-25G 285749-5G
SIAL285749-25GEA 4190 CZK
SIAL285749-25G SIAL285749-5G
DL-Ephedrine hydrochloride, Sigma-Aldrich®
DL-Ephedrine hydrochloride
Vzorec: C₆H₅CH(OH)CH(CH₃)NHCH₃·HCl
CAS číslo: 134-71-4
EINECS: 200-074-6
UN: 1544
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR